Specifications
Avg. Mol. Wt. or Mol. Wt. RangeFe:Cr:Ni; 70:19:11 wt%
Chemical Name or MaterialStainless Steel gauze, Type 304
Health Hazard 1H317-H350-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
View more