Cadmium, plasma standard solution, Specpure™ Cd 10μg/mL
Thermo Scientific Chemicals

Cadmium, plasma standard solution, Specpure™ Cd 10μg/mL

 
Have Questions?
Catalog NumberQuantity
045250.AE
also known as 045250-AE
100 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045250.AE
also known as 045250-AE
Price (USD)
-
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialCadmium, plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H290-H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only