Specifications
Chemical Name or MaterialSelenium, plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1H290-H315-H319
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
View more