Tellurium, plasma standard solution, Specpure™ Te 10μg/mL
Thermo Scientific Chemicals

Tellurium, plasma standard solution, Specpure™ Te 10μg/mL

 
Have Questions?
Catalog NumberQuantity
045265.AE
also known as 045265-AE
100 mL
Catalog number 045265.AE
also known as 045265-AE
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
215.65
Online Exclusive
239.00
Save 23.35 (10%)
Each
Specifications
Chemical Name or MaterialTellurium, plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only