Nickel 61, 61Ni, plasma standard solution, Specpure™, 61Ni 10μg/mL
Thermo Scientific Chemicals

Nickel 61, 61Ni, plasma standard solution, Specpure™, 61Ni 10μg/mL

 
Have Questions?
Catalog NumberQuantity
045282.AD
also known as 045282-AD
50 mL
Catalog number 045282.AD
also known as 045282-AD
Price (USD)
314.65
Special offer
Online exclusive
Ends: 30-Jun-2026
370.00
Save 55.35 (15%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
314.65
Special offer
Online exclusive
Ends: 30-Jun-2026
370.00
Save 55.35 (15%)
Each
Specifications
Chemical Name or MaterialNickel 61, 61Ni, plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1H290-H315-H319
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only