Selenium 82, 82Se, plasma standard solution, Specpure™, 82Se 10μg/mL
Thermo Scientific Chemicals

Selenium 82, 82Se, plasma standard solution, Specpure™, 82Se 10μg/mL

 
Have Questions?
Catalog NumberQuantity
045284.AD
also known as 045284-AD
50 mL
Catalog number 045284.AD
also known as 045284-AD
Price (USD)
286.65
Online Exclusive
318.00
Save 31.35 (10%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (USD)
286.65
Online Exclusive
318.00
Save 31.35 (10%)
Each
Specifications
Chemical Name or MaterialSelenium 82, 82Se, plasma standard solution
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Health Hazard 1H290-H315-H319
Health Hazard 2GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3P234-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P390
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only