Specifications
Chemical Name or MaterialThallium, Oil based standard solution
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.
Health Hazard 3GHS P Statement
P261-P301+P310-P304+P340-P312-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL NumberMFCD01095325
View more