Sulfur in Diesel Fuel standard solution, Specpure™, 25μg/g (0.0025%)
Thermo Scientific Chemicals

Sulfur in Diesel Fuel standard solution, Specpure™, 25μg/g (0.0025%)

 
Have Questions?
Catalog NumberQuantity
045312.AE
also known as 045312-AE
100 mL
Catalog number 045312.AE
also known as 045312-AE
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
111.00
Save 11.35 (10%)
Each
Specifications
Chemical Name or MaterialSulfur in Diesel Fuel standard solution
ColorColorless
EINECS Number270-676-1
Health Hazard 1H227-H351
Health Hazard 2GHS H Statement
H351
Suspected of causing cancer.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only