Sulfur in Isooctane standard solution, Specpure™, 500μg/g (0.0500%)
Thermo Scientific Chemicals

Sulfur in Isooctane standard solution, Specpure™, 500μg/g (0.0500%)

 
Have Questions?
Catalog NumberQuantity
045322.AE
also known as 045322-AE
100 mL
Catalog number 045322.AE
also known as 045322-AE
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
102.65
Online Exclusive
114.00
Save 11.35 (10%)
Each
Specifications
Chemical Name or MaterialSulfur in Isooctane standard solution
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
EINECS Number208-759-1
Health Hazard 1H225-H304-H315-H319-H336
Health Hazard 2GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only