Lead in Isooctane standard solution, Specpure™, 370μg/g(1.0g/gal)
Lead in Isooctane standard solution, Specpure™, 370μg/g(1.0g/gal)
Lead in Isooctane standard solution, Specpure™, 370μg/g(1.0g/gal)
Thermo Scientific Chemicals

Lead in Isooctane standard solution, Specpure™, 370μg/g(1.0g/gal)

CAS: 540-84-1 | C8H18 | 114.232 g/mol
Have Questions?
Catalog NumberQuantity
045325.22
also known as 045325-22
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045325.22
also known as 045325-22
Price (USD)
131.65
Online Exclusive
146.00
Save 14.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
131.65
Online Exclusive
146.00
Save 14.35 (10%)
Each
Specifications
CAS540-84-1
ChEBICHEBI:62805
Chemical Name or MaterialLead in Isooctane standard solution
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
Health Hazard 1H225-H304-H315-H319-H336
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only