Lead in Isooctane standard solution, Specpure™, 0.37μg/g(0.001g/gal), Thermo Scientific Chemicals
Lead in Isooctane standard solution, Specpure™, 0.37μg/g(0.001g/gal), Thermo Scientific Chemicals
Lead in Isooctane standard solution, Specpure™, 0.37μg/g(0.001g/gal), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lead in Isooctane standard solution, Specpure™, 0.37μg/g(0.001g/gal), Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
045331.22
also known as 045331-22
100 g
Catalog number 045331.22
also known as 045331-22
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
115.65
Online Exclusive
128.00
Save 12.35 (10%)
Each
Specifications
CAS540-84-1
ChEBICHEBI:62805
Chemical Name or MaterialLead in Isooctane standard solution
ColorColorless
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only