Specifications
Chemical Name or MaterialLead in Isooctane standards
ColorColorless
DOT InformationTransport Hazard Class: 3; Packing Group: II; Proper Shipping Name: OCTANES
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
View more