Bismuth Internal Standard for method ASTM D5059 Part A and C, Bi @ 0.793g/L
Thermo Scientific Chemicals

Bismuth Internal Standard for method ASTM D5059 Part A and C, Bi @ 0.793g/L

 
Have Questions?
Catalog NumberQuantity
045338.22100 g
Catalog number 045338.22
Price (USD)
148.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
148.00
Each
Specifications
Chemical Name or MaterialBismuth Internal Standard for method ASTM D5059
ColorBrown
Health Hazard 1H335
MDL NumberMFCD00134033
Name NotePart A and C, Bi at 0.793g/L
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only