Sulfur in Light Mineral Oil standard solution, Specpure™, 15μg/g (0.0015%)
Thermo Scientific Chemicals

Sulfur in Light Mineral Oil standard solution, Specpure™, 15μg/g (0.0015%)

 
Have Questions?
Catalog NumberQuantity
045348.AE
also known as 045348-AE
100 mL
Catalog number 045348.AE
also known as 045348-AE
Price (USD)
102.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
102.00
Each
Specifications
Chemical Name or MaterialSulfur in Light Mineral Oil standard solution
MDL NumberMFCD00804270
Name Note15μg/g (0.0015%)
Physical FormLiquid
Quantity100 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only