Sulfur in Heavy Mineral Oil standard solution, Specpure™, 15μg/g (0.0015%)
Thermo Scientific Chemicals

Sulfur in Heavy Mineral Oil standard solution, Specpure™, 15μg/g (0.0015%)

 
Have Questions?
Catalog NumberQuantity
045350.AE
also known as 045350-AE
100 mL
Catalog number 045350.AE
also known as 045350-AE
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
91.65
Online Exclusive
102.00
Save 10.35 (10%)
Each
Specifications
Chemical Name or MaterialSulfur in Heavy Mineral Oil standard solution
Health Hazard 1H304
Health Hazard 3P301+P310-P331-P501c
MDL NumberMFCD00804270
Name Note15μg/g (0.0015%)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only