Cadmium selenide, 99.999% (metals basis)
Cadmium selenide, 99.999% (metals basis)
Cadmium selenide, 99.999% (metals basis)
Thermo Scientific Chemicals

Cadmium selenide, 99.999% (metals basis)

CAS: 1306-24-7 | CdSe | 191.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045456.09
also known as 045456-09
Price (USD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS1306-24-7
IUPAC Nameselanylidenecadmium
Molecular FormulaCdSe
InChI KeyAQCDIIAORKRFCD-UHFFFAOYSA-N
SMILES[Se]=[Cd]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Cadmium selenide is applied in opto-electronic devices, laser diodes, biomedical imaging, nanosensing, high-efficiency solar cells and thin-film transistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only