Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol
Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol
Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol
Thermo Scientific Chemicals

Tin(IV) isopropoxide, 99% (metals basis), 10% w/v in isopropanol

CAS: 1184-61-8 | C12H32O4Sn | 359.094 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
500 mL
Catalog number 045480.AC
also known as 045480-AC
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
126.65
Online Exclusive
140.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS1184-61-8
IUPAC Nametetrakis(propan-2-ol) tin
Molecular FormulaC12H32O4Sn
InChI KeyVUDAJANAMZGVOP-UHFFFAOYSA-N
SMILES[Sn].CC(C)O.CC(C)O.CC(C)O.CC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to red
FormLiquid
Concentration (w/v)9.5-10.5%
Total Metal Impurities≤1%
Tin(IV) isopropoxide is used as a precursor for the preparation of tin oxide nanoparticles, thin films, indium tin oxide (ITO) films , which shows desired electrochemical and conducting properties. Doped tin oxide nanoparticles and thin films find application as gas sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only