Zinc oxide substrate, 10x10x0.5mm, polished one side, 0001 orientation
Zinc oxide substrate, 10x10x0.5mm, polished one side, 0001 orientation
Zinc oxide substrate, 10x10x0.5mm, polished one side, 0001 orientation
Thermo Scientific Chemicals

Zinc oxide substrate, 10x10x0.5mm, polished one side, 0001 orientation

0001 Orientation | CAS: 1314-13-2 | OZn | 81.38 g/mol
Have Questions?
Catalog number 045486.KT
also known as 045486-KT
Price (USD)
600.65
Online Exclusive
667.00
Save 66.35 (10%)
Each
Request bulk or custom format
Price (USD)
600.65
Online Exclusive
667.00
Save 66.35 (10%)
Each
Specifications
CAS1314-13-2
ChEBICHEBI:36560
Chemical Name or MaterialZinc Oxide Substrate
ColorWhite
DOT InformationHazard Class: 9; Packaging Group: III
View more
Zinc oxide substrate is one of the suitable substrate for use as wide band gap semiconductor application. It is also used in power device, LED, sensor and detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only