Methyl cellulose, viscosity 15 cPs
Methyl cellulose, viscosity 15 cPs
Methyl cellulose, viscosity 15 cPs
Thermo Scientific Chemicals

Methyl cellulose, viscosity 15 cPs

CAS: 9004-67-5 | C20H38O11 | 454.513 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 045490.36
also known as 045490-36
Price (USD)
126.00
Each
Quantity:
500 g
Request bulk or custom format
Price (USD)
126.00
Each
Chemical Identifiers
CAS9004-67-5
IUPAC Name3,4,5-trimethoxy-2-(methoxymethyl)-6-{[4,5,6-trimethoxy-2-(methoxymethyl)oxan-3-yl]oxy}oxane
Molecular FormulaC20H38O11
InChI KeyYLGXILFCIXHCMC-UHFFFAOYNA-N
SMILESCOCC1OC(OC2C(COC)OC(OC)C(OC)C2OC)C(OC)C(OC)C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Viscosity12-18 cPs for a 2% aqueous solution at 20?C
Appearance (Color)White to pale yellow
Methyl cellulose is used as an emulsifying and suspending agent in cosmetics, pharmaceutics and the chemical industry. It is used therapeutically as a bulk laxative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only