Nitinol foil, 0.05mm (0.002in) thick, superelastic, flat annealed, pickled surface
Thermo Scientific Chemicals

Nitinol foil, 0.05mm (0.002in) thick, superelastic, flat annealed, pickled surface

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Catalog number 045492.FF
also known as 045492-FF
Price (USD)
77.65
Online Exclusive
86.20
Save 8.55 (10%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
77.65
Online Exclusive
86.20
Save 8.55 (10%)
Each
Specifications
Chemical Name or MaterialNitinol foil
Health Hazard 1Danger
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL NumberMFCD02091734
View more
Nitinol foil is used in cell-phone technology as a retractable antenna, or microphone boom, due to its highly flexible and mechanical memory nature. It is also used in some novelty products, such as self-bending spoons which can be used by amateur and stage magicians to demonstrate psychic powers or as a practical joke, as the spoon will bend itself when used to stir tea, coffee, or any other warm liquid. It is highly biocompatible and has properties suitable for use in orthopaedic implants. Due to Nitinol's unique properties it has seen a large demand for use in less invasive medical devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only