Bismuth 2-ethylhexanoate acid
Bismuth 2-ethylhexanoate acid
Bismuth 2-ethylhexanoate acid
Bismuth 2-ethylhexanoate acid
Thermo Scientific Chemicals

Bismuth 2-ethylhexanoate acid

CAS: 67874-71-9 | C24H48BiO6 | 641.622 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 045500.22
also known as 045500-22
Price (USD)
326.00
Each
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
326.00
Each
Add to cart
Chemical Identifiers
CAS67874-71-9
IUPAC Namebismuth(3+) tris(2-ethylhexanoate)
Molecular FormulaC24H45BiO6
InChI KeyNUMHJBONQMZPBW-UHFFFAOYNA-K
SMILES[Bi+3].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
Assay from Supplier's CofA≥60.0%
FormLiquid to viscous liquid
Bismuth 2-ethylhexanoate is used as an industrial lubricant, dessicant, and polyurethane catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only