Manganese nanopowder, APS 30-50nm
Manganese nanopowder, APS 30-50nm
Manganese nanopowder, APS 30-50nm
Thermo Scientific Chemicals

Manganese nanopowder, APS 30-50nm

Form: Mn3O4 layer 5-12nm | CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 045508.06
also known as 045508-06
Price (USD)
253.65
Special offer
Online exclusive
Ends: 28-Dec-2026
298.00
Save 44.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
253.65
Special offer
Online exclusive
Ends: 28-Dec-2026
298.00
Save 44.35
Each
Specifications
AppearanceGray powder
Assay Percent Range99.9% (Metals basis)
CAS7439-96-5
ChEBICHEBI:35154
Chemical Name or MaterialManganese
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only