Indium(III) fluoride, anhydrous, 99.95% (metals basis)
Indium(III) fluoride, anhydrous, 99.95% (metals basis)
Indium(III) fluoride, anhydrous, 99.95% (metals basis)
Thermo Scientific Chemicals

Indium(III) fluoride, anhydrous, 99.95% (metals basis)

CAS: 7783-52-0 | F3In
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 045525.14
also known as 045525-14
Price (USD)
412.65
Online Exclusive
458.00
Save 45.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
412.65
Online Exclusive
458.00
Save 45.35 (10%)
Each
Chemical Identifiers
CAS7783-52-0
IUPAC Nameindium(3+) trifluoride
Molecular FormulaF3In
InChI KeyJNLSTWIBJFIVHZ-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[In+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder and/or lumps
Total Metal Impurities≤0.05%
Indium(III) fluoride is used in the preparation of non-oxide glasses. It acts as a catalyst in the reaction of trimethylsilyl cyanide (TMSCN) to aldehydes to get cyanohydrins. It play a vital role as a catalyst in organic synthesis to prepare fluorine containing 2,4,5-trisubstituted imidazoles and polysubstituted pyrroles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only