Gold wire, 0.25mm (0.01in) dia, annealed, Premion™, 99.999% (metals basis)
Gold wire, 0.25mm (0.01in) dia, annealed, Premion™, 99.999% (metals basis)
Gold wire, 0.25mm (0.01in) dia, annealed, Premion™, 99.999% (metals basis)
Thermo Scientific Chemicals

Gold wire, 0.25mm (0.01in) dia, annealed, Premion™, 99.999% (metals basis)

CAS: 7440-57-5 | Au | 196.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 cm
100 cm
500 cm
Catalog number 045567.CC
also known as 045567-CC
Price (USD)
-
Quantity:
500 cm
Request bulk or custom format
Specifications
Assay Percent Notes(metals basis)
CAS7440-57-5
ChEBICHEBI:30050
Chemical Name or MaterialGold wire
EINECS Number231-165-9
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only