Strontium perchlorate trihydrate, 98%
Strontium perchlorate trihydrate, 98%
Strontium perchlorate trihydrate, 98%
Thermo Scientific Chemicals

Strontium perchlorate trihydrate, 98%

CAS: 15650-09-6 | Cl2H6O11Sr | 340.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
Catalog number 045577.09
also known as 045577-09
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.20
Save 5.55 (14%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 30-Jun-2026
39.20
Save 5.55 (14%)
Each
Chemical Identifiers
CAS15650-09-6
IUPAC Namestrontium(2+) trihydrate diperchlorate
Molecular FormulaCl2H6O11Sr
InChI KeyVRRVWIQWZWFYDJ-UHFFFAOYSA-L
SMILESO.O.O.[Sr++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)25.2-26.2% Sr
Strontium perchlorate trihydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only