Copper germanium selenide, 99.99% (metals basis), Thermo Scientific Chemicals
Copper germanium selenide, 99.99% (metals basis), Thermo Scientific Chemicals
Copper germanium selenide, 99.99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Copper germanium selenide, 99.99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
045605.09
also known as 045605-09
10 g
Catalog number 045605.09
also known as 045605-09
Price (USD)
209.65
Special offer
Online exclusive
Ends: 28-Dec-2026
246.00
Save 36.35
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
209.65
Special offer
Online exclusive
Ends: 28-Dec-2026
246.00
Save 36.35
Each
Chemical Identifiers
CAS54723-90-9
IUPAC Namecupriogermaneselone
Molecular FormulaCuGeSe
InChI KeyLQTVKWORTHHMFT-UHFFFAOYSA-N
SMILES[Cu]=[Ge]=[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.
The most notable physical characteristics of germania (GeO2) are its high index of refraction and its low optical dispersion. These make it especially useful for wide-angle camera lenses, microscopy, and for the core part of optical fibers. Germanium-on-insulator substrates are seen as a potential replacement for silicon on miniaturized chips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only