Ytterbium sputtering target, 50.8mm dia x 1.59mm thick
Ytterbium sputtering target, 50.8mm dia x 1.59mm thick
Ytterbium sputtering target, 50.8mm dia x 1.59mm thick
Thermo Scientific Chemicals

Ytterbium sputtering target, 50.8mm dia x 1.59mm thick

CAS: 7440-64-4 | Yb | 173.05 g/mol
Have Questions?
Catalog NumberQuantity
045620.KS1 Ea.
Catalog number 045620.KS
Price (USD)
1,057.65
Online Exclusive
1,175.00
Save 117.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
1,057.65
Online Exclusive
1,175.00
Save 117.35 (10%)
Each
Chemical Identifiers
CAS7440-64-4
IUPAC Nameytterbium
Molecular FormulaYb
InChI KeyNAWDYIZEMPQZHO-UHFFFAOYSA-N
SMILES[Yb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Gray
FormSputtering target, 50.8mm diameter x 1.59mm thick.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only