Gadolinium sputtering target, 50.8mm dia x 3.18mm thick
Gadolinium sputtering target, 50.8mm dia x 3.18mm thick
Gadolinium sputtering target, 50.8mm dia x 3.18mm thick
Thermo Scientific Chemicals

Gadolinium sputtering target, 50.8mm dia x 3.18mm thick

CAS: 7440-54-2 | Gd | 157.25 g/mol
Have Questions?
Catalog NumberQuantity
045737.KS1 Ea.
Catalog number 045737.KS
Price (USD)
1,007.65
Online Exclusive
1,119.00
Save 111.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
1,007.65
Online Exclusive
1,119.00
Save 111.35 (10%)
Each
Specifications
CAS7440-54-2
ChEBICHEBI:33375
Chemical Name or MaterialGadolinium sputtering target
EINECS Number231-162-2
IUPAC Namegadolinium
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only