Copper tubing, 6.35mm (0.25in) OD, 4.72mm (0.186in) ID
Copper tubing, 6.35mm (0.25in) OD, 4.72mm (0.186in) ID
Copper tubing, 6.35mm (0.25in) OD, 4.72mm (0.186in) ID
Thermo Scientific Chemicals

Copper tubing, 6.35mm (0.25in) OD, 4.72mm (0.186in) ID

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Catalog NumberQuantity
045793.N610 ft.
Catalog number 045793.N6
Price (USD)
86.65
Online Exclusive
96.10
Save 9.45 (10%)
Each
Quantity:
10 ft.
Request bulk or custom format
Price (USD)
86.65
Online Exclusive
96.10
Save 9.45 (10%)
Each
Specifications
CAS7440-50-8
ChEBICHEBI:30052
Chemical Name or MaterialCopper tubing
EINECS Number231-159-6
IUPAC Namecopper
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only