Silica gel, HPLC grade, spherical, 3 micron APS, 120 angstroms, 99.99+%
Silica gel, HPLC grade, spherical, 3 micron APS, 120 angstroms, 99.99+%
Silica gel, HPLC grade, spherical, 3 micron APS, 120 angstroms, 99.99+%
Thermo Scientific Chemicals

Silica gel, HPLC grade, spherical, 3 micron APS, 120 angstroms, 99.99+%

CAS: 63231-67-4 | H2O3Si | 78.098 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 045803.03
also known as 045803-03
Price (USD)
58.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
58.10
Each
Specifications
CAS63231-67-4
Chemical Name or MaterialSilica gel
EINECS Number231-545-4
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H373-H319-H335
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only