Cobalt(II,III) oxide, 99.7% (metals basis)
Cobalt(II,III) oxide, 99.7% (metals basis)
Cobalt(II,III) oxide, 99.7% (metals basis)
Cobalt(II,III) oxide, 99.7% (metals basis)
Thermo Scientific Chemicals

Cobalt(II,III) oxide, 99.7% (metals basis)

CAS: 1308-06-1 | Co3O4 | 240.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1000 g
Catalog number 045806.18
also known as 045806-18
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS1308-06-1
IUPAC Namedicobalt(3+) λ²-cobalt(2+) tetraoxidandiide
Molecular FormulaCo3O4
InChI KeyUBEWDCMIDFGDOO-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[O--].[Co++].[Co+3].[Co+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.3% max.

Glassy carbon is widely practiced as an electrode material in electrochemistry, as considerably as for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only