Silica gel, HPLC grade, spherical, 5 micron APS, 70 angstroms, 99.99+%
Thermo Scientific Chemicals

Silica gel, HPLC grade, spherical, 5 micron APS, 70 angstroms, 99.99+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 045809.14
also known as 045809-14
Price (USD)
514.65
Online Exclusive
572.00
Save 57.35 (10%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
514.65
Online Exclusive
572.00
Save 57.35 (10%)
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentPore diameter 6.0-9.0 nm
CommentPore volume 0.85-1.05 ml/g
Particle Size4.30-4.50 Microns
Surface Area460-540 m²/g
pH4.0 min
View more
Silica gel, HPLC grade is used in chromatography applications, separation and purification. It is also used in preparative chromatography for HPLC packing, catalyst carriers and resin fillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only