Silica gel, preparative chromatography grade, spherical, 10 micron APS, 60 angstroms, 99.99+%
Silica gel, preparative chromatography grade, spherical, 10 micron APS, 60 angstroms, 99.99+%
Silica gel, preparative chromatography grade, spherical, 10 micron APS, 60 angstroms, 99.99+%
Thermo Scientific Chemicals

Silica gel, preparative chromatography grade, spherical, 10 micron APS, 60 angstroms, 99.99+%

CAS: 63231-67-4 | H2O3Si | 78.098 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 045819.06
also known as 045819-06
Price (USD)
51.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
51.00
Each
Chemical Identifiers
CAS7631-86-9
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, spherical
Assay from Suppliers CofA≥99.99%
Particle SizeAPS: 9.0-11.0 µm
Particle SizePore diameter: 4.8-7.8 nm (~60 angstroms)
Particle SizePore Volume: 0.8-1.2 mL/g
View more
Silica gel is mainly used for the dehydrated purification of the industrial gases, the clearance of the organic acids and high polymers in the insulative oil, the purification and separation of the medicines. It is also used in column chromatography without applied pressure (gravity pulled).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only