Zeolite Y, ammonium
Zeolite Y, ammonium
Zeolite Y, ammonium
Thermo Scientific Chemicals

Zeolite Y, ammonium

Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. Used in detergent manufacturing for the applications of the removal of hardness. | CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 045863.14
also known as 045863-14
Price (USD)
32.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
32.00
Each
Chemical Identifiers
CAS1318-02-1
IUPAC Nameoxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular FormulaAl2O5Si
InChI KeyHNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILESO=[Si]=O.O=[Al]O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSurface area 925 m²/g
CommentSiO₂:Al₂O₃ mole ratio 5.1:1
Zeolite Y is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only