Zeolite Y, hydrogen
Zeolite Y, hydrogen
Zeolite Y, hydrogen
Thermo Scientific Chemicals

Zeolite Y, hydrogen

Zeolite Y is a useful metal aluminum silicate for proteomics research. It is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. | CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 045871.22
also known as 045871-22
Price (USD)
86.40
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
86.40
Each
Chemical Identifiers
CAS1318-02-1
IUPAC Nameoxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular FormulaAl2O5Si
InChI KeyHNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILESO=[Si]=O.O=[Al]O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
CommentSiO₂:Al₂O₃ molar ratio 45 - 72
CommentSurface area >650 m²/g
Zeolite Y is a useful metal aluminum silicate for proteomics research. It is used as a cracking catalyst in petroleum refinery catalytic cracking units to convert high-boiling fractions of petroleum crude to more valuable gasoline, diesel and other products. It is also used as a molecular sieve and as an adsorbent of a wide variety of cations, such as Na+, K+, Ca2+, Mg2+ and others. Zeolite is used in the development of a variety of bioreactor devices and in the determination of serum triglycerides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only