Zeolite ZSM-5, ammonium
Zeolite ZSM-5, ammonium
Zeolite ZSM-5, ammonium
Thermo Scientific Chemicals

Zeolite ZSM-5, ammonium

CAS: 1318-02-1 | Al2O5Si | 162.043 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number 045880.22
also known as 045880-22
Price (USD)
91.60
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
91.60
Each
Chemical Identifiers
CAS1318-02-1
IUPAC Nameoxo[(oxoalumanyl)oxy]alumane; silanedione
Molecular FormulaAl2O5Si
InChI KeyHNPSIPDUKPIQMN-UHFFFAOYSA-N
SMILESO=[Si]=O.O=[Al]O[Al]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)SiO₂:Al₂O₃ mole ratio 27-34:1
Assay (unspecified)Surface area 390-450 m²/g
Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support. Used in detergent manufacturing for the applications of the removal of hardness. It is also used as Odor control, industrial absorbents, industrial fillers, gas absorption, soil remediation, wastewater filtration, flocculating agent, animal feeds, hydroponics, molecular sieves, desiccants, lightweight concrete, acoustics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zeolite ZSM-5 is used in hydrocarbon isomerization, the alkylation of hydrocarbons, and as a catalyst support
RUO – Research Use Only