How many Aluminum (Al) atoms are present in each molecule of Zeolite ZSM-5, ammonium?
The molecular formula of Zeolite ZSM-5, ammonium is Al2O5Si, which indicates that there are 2 aluminum (Al) atoms in each molecule.
Search
View additional product information for Zeolite ZSM-5, ammonium - FAQs (045882.22, 045882.14, 045882.36)
1 product FAQs found