1L Solvent Bottle for Dionex™ ASE™ and AutoTrace™ Systems
Thermo Scientific™
1L Solvent Bottle for Dionex™ ASE™ and AutoTrace™ Systems
Use this 1L bottle to hold solvent on the Thermo Scientific™ Dionex™ ASE™ Accelerated Solvent Extractor or AutoTrace™ Solid Phase Extraction (SPE) systems.
Have Questions?
Catalog NumberVolume (Metric)
0459001 L
Catalog number 045900
Price (USD)
129.00
Each
Volume (Metric):
1 L
Price (USD)
129.00
Each
Use this 1L bottle to hold solvent on the Thermo Scientific™ Dionex™ ASE™ 150/350 Accelerated Solvent Extractor or Thermo Scientific™ Dionex™ AutoTrace™ 280 Solid Phase Extraction (SPE) systems.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionSolvent Bottle
MaterialGL45 Coated Glass
QuantityEach 1
Volume (Metric)1 L
For Use With (Equipment)Dionex ASE 150, 350; AutoTrace Systems
TypeSolvent Bottle
Unit SizeEach