Ytterbium powder, -200 mesh, 99.9% (REO)
Ytterbium powder, -200 mesh, 99.9% (REO)
Ytterbium powder, -200 mesh, 99.9% (REO)
Thermo Scientific Chemicals

Ytterbium powder, -200 mesh, 99.9% (REO)

CAS: 7440-64-4 | Yb | 173.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 045920.04
also known as 045920-04
Price (USD)
130.65
Special offer
Online exclusive
Ends: 28-Dec-2026
154.00
Save 23.35
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 28-Dec-2026
154.00
Save 23.35
Each
Chemical Identifiers
CAS7440-64-4
IUPAC Nameytterbium
Molecular FormulaYb
InChI KeyNAWDYIZEMPQZHO-UHFFFAOYSA-N
SMILES[Yb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
RUO – Research Use Only