Antimony(III) telluride, 99.999% (metals basis)
Antimony(III) telluride, 99.999% (metals basis)
Antimony(III) telluride, 99.999% (metals basis)
Thermo Scientific Chemicals

Antimony(III) telluride, 99.999% (metals basis)

CAS: 1327-50-0 | Sb2Te3 | 626.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 045922.18
also known as 045922-18
Price (USD)
689.65
Special offer
Online exclusive
Ends: 30-Jun-2026
811.00
Save 121.35 (15%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
689.65
Special offer
Online exclusive
Ends: 30-Jun-2026
811.00
Save 121.35 (15%)
Each
Chemical Identifiers
CAS1327-50-0
IUPAC Namebis(tellanylidenestibanyl) tellurium
Molecular FormulaSb2Te3
InChI KeyBPDQXJZWVBPDSN-UHFFFAOYSA-N
SMILES[Te].[Sb]=[Te].[Sb]=[Te]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Antimony(III) telluride is used as both n-type and p-type semiconductors in association with appropriate dopant. It reacts with germanium telluride and forms the pseudobinary intermetallic system. It is used in solid state refrigerators due to its large thermoelectric effect. Further, it is an active component of thermoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only