Sodium chloride crystal optic rectangle, 41mm x 23mm x 6mm (drilled), polished both sides
Sodium chloride crystal optic rectangle, 41mm x 23mm x 6mm (drilled), polished both sides
Sodium chloride crystal optic rectangle, 41mm x 23mm x 6mm (drilled), polished both sides
Thermo Scientific Chemicals

Sodium chloride crystal optic rectangle, 41mm x 23mm x 6mm (drilled), polished both sides

CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Catalog NumberQuantity
045944.KT
also known as 045944-KT
1 Ea.
Catalog number 045944.KT
also known as 045944-KT
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)
111.65
Online Exclusive
124.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Width23mm
FormCrystal optic rectangle (drilled), polished both sides.
Length41mm
Height6mm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only