Aluminum oxide, basic,  HPLC Flash Grade
Aluminum oxide, basic,  HPLC Flash Grade
Aluminum oxide, basic,  HPLC Flash Grade
Thermo Scientific Chemicals

Aluminum oxide, basic, HPLC Flash Grade

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 045949.30
also known as 045949-30
Price (USD)
-
Quantity:
250 g
Request bulk or custom format
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
pH9.0-10.0
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only