Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0), Pd 16.7%
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0), Pd 16.7%
Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0), Pd 16.7%
Thermo Scientific Chemicals

Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0), Pd 16.7%

CAS: 1233717-68-4 | C32H56N2P2Pd | 637.182 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number 046000.03
also known as 046000-03
Price (USD)
495.65
Online Exclusive
550.00
Save 54.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
495.65
Online Exclusive
550.00
Save 54.35 (10%)
Each
Chemical Identifiers
CAS1233717-68-4
IUPAC Namebis(4-(di-tert-butylphosphanyl)-N,N-dimethylaniline) palladium
Molecular FormulaC32H56N2P2Pd
InChI KeySSPOQURGNAWORH-UHFFFAOYSA-N
SMILES[Pd].CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Pd 16.7% min

Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(0) is used in cross-coupling including sp2-sp3 coupling, especially Suzuki, Negishi-type, Heck, Sonogashira, and Buchwald-Hartwig coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only