Dionex™ IonPac™ AS12A IC Columns
Actual product may vary
Dionex™ IonPac™ AS12A IC Columns
Dionex™ IonPac™ AS12A IC Columns
Thermo Scientific™

Dionex™ IonPac™ AS12A IC Columns

Green features

Medium capacity anion-exchange column using a carbonate/bicarbonate eluent for separating inorganic anions and oxyhalides, such as bromate, in environmental waters.

Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Column Type
046034200 mm4 mmAnalytical Column
046055200 mm2 mmAnalytical Column
04605650 mm2 mmGuard Column
07980150 mm4 mmGuard Column
Catalog number 046034
Price (USD)
1,930.00
Each
Length (Metric):
200 mm
Diameter (Metric):
4 mm
Column Type:
Analytical Column
Price (USD)
1,930.00
Each

Separate inorganic anions and oxyhalides, such as chlorite and bromate, in environmental samples. The Thermo Scientific™ Dionex™ IonPac™ AS12A Analytical and Guard Columns resolve trace chloride and sulfate in high-carbonate samples and meet or exceed the requirements of U.S EPA Methods 300.0 (A) and (B). However, the advanced, high-capacity IonPac AS23 Carbonate Eluent Anion-Exchange column is recommended for determination of oxyhalides and inorganic anions, such as trace bromate, in ozonated drinking water.

Fast Isocratic Separation of Inorganic Anions and Oxyhalides in Diverse Matrices
  • Compliance monitoring of common anions in drinking water as specified in U.S. EPA Method 300.0 (A) and (B)
  • Separation of common inorganic anions and oxyhalides including chlorite, bromate and chlorate in less than 12 minutes
  • Superior retention and quantification of fluoride
  • Trace analysis of chloride and sulfate in high-carbonate samples
  • Solvent compatibility permits easy column cleanup after the analysis of samples with hydrophobic components.

Multiple Formats Available for Application Flexibility
  • 4x200mm, 2x200mm analytical columns
  • 4x50mm, 2x50mm guard columns
  • Longer 200mm columns facilitate higher resolution separations of more complex sample matrices
  • Smaller i.d. columns provide higher mass sensitivity, eluent and reagent conservation and lower operating costs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Anion Exchange Capacity52 μeq
Certifications/ComplianceU.S. EPA Methods 300.0 (A) and (B)
Column Capacity52 μeq
Column ChemistryCarbonate/Bicarbonate
Column ConstructionPEEK
Crosslinking55% DVB
Diameter (Metric) Latex140 nm
Flow Rate1.5 to 2.0 mL/min.
For Use With (Application)Separation of Inorganic Anions
For Use With (Equipment)ASRS 300 System
Green FeaturesFSC
HydrophobicityMedium
Ion Exchange GroupAlkyl Quaternary Ammonium
Max. Pressure4000 psi (275 bar)
Packing MaterialEthylvinylbenzene/Divinylbenzene
Particle Size9 μm
Pore Size2000 &‌Aring;
QuantityEach 1
Solvent CompatibilitypH 0 to 14, 0 to 100% HPLC Solvents
Stationary PhaseIonPac AS12A
Column TypeAnalytical Column
Diameter (Metric)4 mm
FormatStandard Bore
Length (Metric)200 mm
Unit SizeEach