Aluminum oxide, basic, HPLC Flash Grade
Aluminum oxide, basic, HPLC Flash Grade
Aluminum oxide, basic, HPLC Flash Grade
Thermo Scientific Chemicals

Aluminum oxide, basic, HPLC Flash Grade

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 046071.36
also known as 046071-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point2980°C
CAS1344-28-1
Chemical Name or MaterialAluminum oxide
ColorWhite
EINECS Number215-691-6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only