n-Butyl acetate, ACS Grade, 99.5+%
Thermo Scientific™

n-Butyl acetate, ACS Grade, 99.5+%

CAS: 123-86-4 | C6H12O2 | 116.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 046186.K7
also known as 046186-K7
Price (USD)
273.00
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
273.00
Each
Chemical Identifiers
CAS123-86-4
IUPAC Namebutyl acetate
Molecular FormulaC6H12O2
InChI KeyDKPFZGUDAPQIHT-UHFFFAOYSA-N
SMILESCCCCOC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay (GC)≥99.5%
Free acid (titration)Titrable acid: ≤0.0016 meq/g
View more