Platinum, 5% on activated carbon powder, Type B105022-5, standard, reduced, nominally 50% water wet
Platinum, 5% on activated carbon powder, Type B105022-5, standard, reduced, nominally 50% water wet
Platinum, 5% on activated carbon powder, Type B105022-5, standard, reduced, nominally 50% water wet
Thermo Scientific Chemicals

Platinum, 5% on activated carbon powder, Type B105022-5, standard, reduced, nominally 50% water wet

CAS: 7440-6-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 046306.06
also known as 046306-06
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Specifications
CAS6-6-7440
Chemical Name or MaterialPlatinum
ColorGray to Black
MDL NumberMFCD00011179
Name Note5% on activated carbon powder, Type B105022-5, Standard, Reduced, Nominally 50% water wet
View more

Platinum, 5% on activated carbon powder, Type B105022-5 is used as a catalyst for the hydrogenation of alkenes, alkynes, nitro groups, ketones, and aromatics. It is also used for hydrogenolysis of cyclopropanes. Further, it is involved in the oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only