Silicon powder, crystalline, APS 100nm, 99%, Plasma Synthesized
Silicon powder, crystalline, APS 100nm, 99%, Plasma Synthesized
Silicon powder, crystalline, APS 100nm, 99%, Plasma Synthesized
Silicon powder, crystalline, APS 100nm, 99%, Plasma Synthesized
Thermo Scientific Chemicals

Silicon powder, crystalline, APS 100nm, 99%, Plasma Synthesized

CAS: 7440-21-3 | Si | 28.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 046308.04
also known as 046308-04
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.80
Save 8.15 (14%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 30-Jun-2026
57.80
Save 8.15 (14%)
Each
Chemical Identifiers
CAS7440-21-3
IUPAC Namesilicon
Molecular FormulaSi
InChI KeyXUIMIQQOPSSXEZ-UHFFFAOYSA-N
SMILES[Si]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)99%
Particle SizeAPS ≈100nm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only