Fullerene powder, 99% C{6}{0}
Fullerene powder, 99% C{6}{0}
Fullerene powder, 99% C{6}{0}
Fullerene powder, 99% C{6}{0}
Thermo Scientific Chemicals

Fullerene powder, 99% C{6}{0}

CAS: 99685-96-8 | C60 | 720.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number 046311.03
also known as 046311-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS99685-96-8
IUPAC Name(C60-Ih)[5,6]fullerene
Molecular FormulaC60
InChI KeyXMWRBQBLMFGWIX-UHFFFAOYSA-N
SMILESC12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Assay (HPLC)> 98.5%
Formpowder
It is used in optical computing, optical memories, optical signal processing and controlling applications respects, C60 and its derivatives can be widely used in photovoltaic applications, magnetic resonance imaging, anti-HIV drugs, anti-cancer drugs, chemotherapy drugs, cosmetic additives, research and other areas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only