Potassium hydroxide, 25% w/v aq. soln.
Potassium hydroxide, 25% w/v aq. soln.
Potassium hydroxide, 25% w/v aq. soln.
Thermo Scientific Chemicals

Potassium hydroxide, 25% w/v aq. soln.

CAS: 1310-58-3 | HKO | 56.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 046330.AP
also known as 046330-AP
Price (USD)
44.70
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
44.70
Each
Chemical Identifiers
CAS1310-58-3
IUPAC Namepotassium hydroxide
Molecular FormulaHKO
InChI KeyKWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES[OH-].[K+]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥23.75 to ≤26.25%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only